Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTF2E1, Mouse, Clone: 1E12, Abnova™

Catalog No. 89104602 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-602 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-602 Supplier Abnova Corporation Supplier No. H00002960M07A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GTF2E1.

Sequence: SCVKEEDMLELLKFDRKQLRSVLNNLKGDKFIKCRMRVETAADGKTTRHNYYFINYRTLVNVVKYKLDHMRRRIETDERDSTNRASFKCPVCSSTFTDLE

Specifications

Antigen GTF2E1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GTF2E1.
Formulation ascites with no preservative
Gene GTF2E1
Gene Accession No. NM_005513
Gene Alias FE/TF2E1/TFIIE-A
Gene Symbols GTF2E1
Host Species Mouse
Immunogen GTF2E1 (NP_005504,41 a.a. ∽ 140 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2960
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.