Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GUCY1B3, Mouse, Clone: GC10, Abnova™

Catalog No. 89111217 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-217 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-217 Supplier Abnova Corporation Supplier No. MAB3033

Mouse monoclonal antibody raised against recombinant GUCY1B3.

Soluble guanylate cyclase (sGC), a heterodimeric protein consisting of an alpha subunit and a beta subunit, typically GUCY1B3, catalyzes conversion of GTP to the second messenger cGMP and functions as the main receptor for nitric oxide (NO) and nitrovasodilator drugs (Zabel et al., 1998 [PubMed 9742212]).[supplied by OMIM

Specifications

Antigen GUCY1B3
Applications ELISA, Western Blot
Classification Monoclonal
Clone GC10
Conjugate Unconjugated
Description guanylate cyclase 1, soluble, beta 3
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene GUCY1B3
Gene Alias GC-S-beta-1, GC-SB3, GUC1B3, GUCB3, GUCSB3, GUCY1B1
Gene Symbols GUCY1B3
Host Species Mouse
Immunogen Recombinant protein corresponding to human GUCY1B3.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2983
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.