Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GYPA, Mouse, PE, Clone: HIR2, Abnova™

Catalog No. 89111799 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-799 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-799 Supplier Abnova Corporation Supplier No. MAB4393
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of GYPA.

Glycophorins A (GYPA) and B (GYPB) are major sialoglycoproteins of the human erythrocyte membrane which bear the antigenic determinants for the MN and Ss blood groups. In addition to the M or N and S or s antigens that commonly occur in all populations, about 40 related variant phenotypes have been identified. These variants include all the variants of the Miltenberger complex and several isoforms of Sta, as well as Dantu, Sat, He, Mg, and deletion variants Ena, S-s-U- and Mk. Most of the variants are the result of gene recombinations between GYPA and GYPB. [provided by RefSeq

Specifications

Antigen GYPA
Applications Agglutination, Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone HIR2
Conjugate PE
Description Mouse monoclonal antibody raised against synthetic peptide of GYPA.
Dilution Flow Cytometry (20 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene GYPA
Gene Alias CD235a/GPA/GPErik/GPSAT/GpMiIII/HGpMiIII/HGpMiV/HGpMiX/HGpMiXI/HGpSta(C)/MN/MNS
Gene Symbols GYPA
Host Species Mouse
Immunogen A synthetic peptide corresponding to N-terminus of human GYPA.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2993
Test Specificity This antibody recognizes N-terminal portion of glycophorin A (and weakly of glycophorin B). Its antigen is expressed on early erythroblasts, late erythroblasts, erythroblasts, mature erythrocytes and the cells of erythroid cell lines K562 and HEL, but not on all other cells.
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.