Learn More
GYS1, Mouse, Clone: 3A7, Abnova™
Mouse monoclonal antibody raised against partial recombinant GYS1.
Supplier: Abnova Corporation MAB10677
Description
The protein encoded by this gene catalyzes the addition of glucose monomers to the growing glycogen molecule through the formation of alpha-1,4-glycoside linkages. Mutations in this gene are associated with muscle glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene
Specifications
GYS1 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. | |
GYS1 | |
GYS1 | |
Recombinant protein corresponding to human GYS1. | |
RUO | |
2997 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
3A7 | |
Mouse monoclonal antibody raised against partial recombinant GYS1. | |
In ascites (0.03% sodium azide) | |
GSY/GYS | |
Mouse | |
100 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.