Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H2-D4, Mouse, Clone: 34-5-8S, Abnova™

Catalog No. 89135416 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-416 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-416 Supplier Abnova Corporation Supplier No. MAB3260
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against H2-D4.

Specifications

Antigen H2-D4
Applications Flow Cytometry
Classification Monoclonal
Clone 34-5-8S
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against H2-D4.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Host Species Mouse
Immunogen C57BL/6 x DBA/2)F1 hybrid mouse splenocytes.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity A conformational epitope on the alpha1 and alpha2 chains of mouse H-2Dd class I alloantigen.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.