Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H3F3A/H3F3B, Mouse, Clone: 44AT1232.94, Abnova™

Catalog No. 89134878 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-878 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-878 Supplier Abnova Corporation Supplier No. MAB1149

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse monoclonal antibody raised against synthetic phosphopeptide of H3F3A/H3F3B.

Specifications

Antigen H3F3A/H3F3B
Applications ELISA, Western Blot
Classification Monoclonal
Clone 44AT1232.94
Conjugate Unconjugated
Formulation In PBS (0.09% sodium azide)
Host Species Mouse
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to amino acids 7-18 residues surrounding S10 of human H3F3A/H3F3B.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.