Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HA (A/Chicken/Hong Kong/G9/97)(H9N2), Mouse, Clone: 355, Abnova™

Catalog No. 89134780 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-780 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-780 Supplier Abnova Corporation Supplier No. MAB10126

Mouse monoclonal antibody raised against recombinant HA (H9N2).

Specifications

Antigen HA (A/Chicken/Hong Kong/G9/97)(H9N2)
Applications Dot Blot, ELISA, Hemagglutination Assay, Western Blot
Classification Monoclonal
Clone 355
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant HA (H9N2).
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Host Species Mouse
Immunogen Recombinant protein corresponding to HA (A/Chicken/Hong Kong/G9/97)(H9N2).
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.