Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haemagglutinin H1, Mouse, Clone: InA97, Abnova™

Catalog No. 89135347 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-347 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-347 Supplier Abnova Corporation Supplier No. MAB3132
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Influenza A virus Haemagglutinin H1.

Specifications

Antigen Haemagglutinin H1
Applications ELISA, Western Blot
Classification Monoclonal
Clone InA97
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Influenza A virus Haemagglutinin H1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Influenza A/NewCaledonia/20/99 (strain H1N1).
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.