Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Anti-Hamster IgG PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems F0120
This item is not returnable.
View return policy
Description
IgG Monoclonal specifically detects IgG in Hamster samples. It is validated for Flow Cytometry.Specifications
IgG | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
IGHG1 | |
Armenian hamster IgG | |
100 Tests | |
Secondary | |
Detects IgG from Armenian Hamster. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Flow Cytometry | |
MAH1.12 | |
Flow Cytometry | |
Immunoglobulin G, ImmunoglobulinG | |
Mouse | |
Affinity Purified | |
RUO | |
3500 | |
Hamster | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction