Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HAPLN2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107249 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-249 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-249 Supplier Abnova Corporation Supplier No. H00060484A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant HAPLN2.

Sequence: HPGPHYLLPPIHEVIHSHRGATATLPCVLGTTPPSYKVRWSKVEPGELRETLILITNGLHARGYGPLGGRARMRRGHRLDASLVIAGVRLEDEGRYRCELINGIEDESVALT

Specifications

Antigen HAPLN2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant HAPLN2.
Formulation 50% glycerol
Gene HAPLN2
Gene Accession No. NM_021817
Gene Alias BRAL1
Gene Symbols HAPLN2
Host Species Mouse
Immunogen HAPLN2 (NP_068589, 31 a.a. ∽ 142 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60484
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.