Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HbA1c, Mouse, Clone: 1D3, Abnova™

Catalog No. 89107823 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-823 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-823 Supplier Abnova Corporation Supplier No. MAB0030M06A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against glycosylated hemoglobin.

The human alpha globin gene cluster located on chromosome 16 spans about 30kb and includes seven loci: 5'- zeta - pseudozeta - mu - pseudoalpha-1 - alpha-2 - alpha-1 - theta - 3'. The alpha-2 (HBA2) and alpha-1 (HBA1) coding sequences are identical. These genes differ slightly over the 5' untranslated regions and the introns, but they differ significantly over the 3' untranslated regions. Two alpha chains plus two beta chains constitute HbA, which in normal adult life comprises about 97% of the total hemoglobin; alpha chains combine with delta chains to constitute HbA-2, which with HbF (fetal hemoglobin) makes up the remaining 3% of adult hemoglobin. Alpha thalassemias result from deletions of each of the alpha genes as well as deletions of both HBA2 and HBA1; some nondeletion alpha thalassemias have also been reported. [provided by RefSeq]

Specifications

Antigen HbA1c
Applications ELISA
Classification Monoclonal
Clone 1D3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against glycosylated hemoglobin.
Formulation In ascites fluid
Gene HBA1
Gene Accession No. NM_000558
Gene Alias HBH/HBA-T3
Gene Symbols HBA1
Host Species Mouse
Immunogen A peptide (conjugated with KLH) corresponding to N-terminus glycosylated HbA1c.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3039
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.