Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HbA1c, Mouse, Clone: GHb2, Abnova™

Catalog No. 89111173 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-173 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-173 Supplier Abnova Corporation Supplier No. MAB2953

Mouse monoclonal antibody raised against glycated (in vitro) synthetic peptide corresponding to the N-terminal end of beta-globin.

The human alpha globin gene cluster located on chromosome 16 spans about 30kb and includes seven loci: 5'- zeta - pseudozeta - mu - pseudoalpha-1 - alpha-2 - alpha-1 - theta - 3'. The alpha-2 (HBA2) and alpha-1 (HBA1) coding sequences are identical. These genes differ slightly over the 5' untranslated regions and the introns, but they differ significantly over the 3' untranslated regions. Two alpha chains plus two beta chains constitute HbA, which in normal adult life comprises about 97% of the total hemoglobin; alpha chains combine with delta chains to constitute HbA-2, which with HbF (fetal hemoglobin) makes up the remaining 3% of adult hemoglobin. Alpha thalassemias result from deletions of each of the alpha genes as well as deletions of both HBA2 and HBA1; some nondeletion alpha thalassemias have also been reported. [provided by RefSeq]

Specifications

Antigen HbA1c
Applications ELISA
Classification Monoclonal
Clone GHb2
Conjugate Unconjugated
Description hemoglobin, alpha 1
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene HBA1
Gene Alias HBH, HBA-T3
Gene Symbols HBA1
Host Species Mouse
Immunogen A synthetic peptide corresponding to N-terminus end of human HBA1.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3039
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.