Learn More
HBB, Mouse, Clone: 99g8, Abnova™
Mouse monoclonal antibody raised against full-length recombinant HBB.
Supplier: Abnova Corporation MAB8859
Description
The alpha (HBA) and beta (HBB) loci determine the structure of the 2 types of polypeptide chains in adult hemoglobin, Hb A. The normal adult hemoglobin tetramer consists of two alpha chains and two beta chains. Mutant beta globin causes sickle cell anemia. Absence of beta chain causes beta-zero-thalassemia. Reduced amounts of detectable beta globin causes beta-plus-thalassemia. The order of the genes in the beta-globin cluster is 5'-epsilon -- gamma-G -- gamma-A -- delta -- beta--3'. [provided by RefSeq]
Specifications
HBB | |
Monoclonal | |
1 ug/uL | |
Mouse monoclonal antibody raised against full length recombinant HBB. | |
In citrate-Tris-HCl, pH7.0 (0.02% Proclin 300) | |
CD113t-C | |
Mouse | |
Affinity purification | |
RUO | |
3043 | |
Human | |
Liquid |
ELISA, Western Blot | |
99g8 | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
HBB | |
HBB | |
Recombinant protein corresponding to full length of human HBB. | |
100 μL | |
Primary | |
This antibody recognizes recombinant protein HBB. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.