Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBEGF, Mouse, Clone: 4G10, Abnova™

Catalog No. 89113022 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-022 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-022 Supplier Abnova Corporation Supplier No. MAB6756

Mouse monoclonal antibody raised against recombinant HBEGF.

Specifications

Antigen HBEGF
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4G10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant HBEGF.
Dilution Western Blot (0.2-1 ug/mL) Immunofluorescence (5∼10 ug/mL) Immunoprecipitation (2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, azide free)
Gene HBEGF
Gene Alias DTR/DTS/DTSF/HEGFL
Gene Symbols HBEGF
Host Species Mouse
Immunogen Recombinant protein corresponding to human HBEGF.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1839
Target Species Human
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.