Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HBVgp2, Mouse, Clone: 4v67, Abnova™

Catalog No. 89113639 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-639 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-113-639 Supplier Abnova Corporation Supplier No. MAB8889
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse monoclonal antibody raised against full-length recombinant HBVgp2.

Specifications

Antigen HBVgp2
Applications ELISA
Classification Monoclonal
Clone 4v67
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant HBVgp2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In citrate-Tris-HCl, pH7.0 (0.02% Proclin 300)
Gene HBVgp2
Gene Symbols HBVgp2
Host Species Mouse
Immunogen Recombinant protein corresponding to full length of HBVgp2.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 944569
Test Specificity This antibody recognizes recombinant protein Hepatitis B virus Pres1. The antibody does not cross-react with Hepatitis B virus Pres2.
Target Species Virus
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.