Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hcn4, Mouse, Clone: S114-10, Abnova™

Catalog No. 89112993 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-993 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-993 Supplier Abnova Corporation Supplier No. MAB6667
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant Hcn4.

Specifications

Antigen Hcn4
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone S114-10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Hcn4.
Dilution Western Blot (1-10 ug/mL) Immunohistochemistry (0.1-1.0 ug/mL) Immunocytochemistry (0.1-1.0 ug/mL) Immunofluorescence (1.0-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Hcn4
Gene Symbols Hcn4
Host Species Mouse
Immunogen Recombinant fusion protein corresponding to C-terminus amino acids 1019-1198 of rat Hcn4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59266
Test Specificity Detects ∼130KDa. No cross-reactivity against other HCNs.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.