Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Mouse anti-HDAC1, Clone: 2E10,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ 05100I
Description
Specifically detects HDAC1 clone: 2E10 in Human, Mouse samples, and it is validated for Immunoprecipitation, Western Blotting
Histone deacetylase 1 is a nuclear enzyme encoded by the Hdac1 gene (also known as HD1, RPD3, MommeD5, Hdac1-ps) in mouse and HDAC1 (also known as GoON-10, RPD3L1) gene in human. Hdac1 plays an important role in epigenetic regulation of gene expression by catalyzing the hydrolysis of N(6)-acetylated histone lysine residues. Hdac1 is also known to deacetylate p53 and thereby downregulate p53 activity.
Specifications
HDAC1 | |
Monoclonal | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Hdac1 | |
KLH-conjugated, synthetic peptide (C-EEKPEAKGVKEEVKLA) corresponding to amino acids 467-482 of mouse HDAC1. The immunizing sequence is identical in human. | |
100 μL | |
Epigenetics and Nuclear Function | |
NP_032254 | |
Stable for one year at 2°C to 8°C from date of receipt. | |
IgG1 κ |
Immunoprecipitation, Western Blot | |
2E10 | |
O09106 | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction