Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis A virus, Mouse, Clone: 581, Abnova™

Catalog No. 89135746 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-746 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-746 Supplier Abnova Corporation Supplier No. MAB4726
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against hepatitis A virus.

Specifications

Antigen Hepatitis A virus
Applications ELISA
Classification Monoclonal
Clone 581
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against hepatitis A virus.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (0.09% sodium azide)
Host Species Mouse
Immunogen Lysate of RMK cells infected with an isolate of hepatitis A virus.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the surface protein of Hepatitis A Virus.
Target Species Virus
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.