Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis A virus, Mouse, Clone: 818, Abnova™

Catalog No. 89135747 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-747 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-747 Supplier Abnova Corporation Supplier No. MAB4727

Mouse monoclonal antibody raised against hepatitis A virus.

Specifications

Antigen Hepatitis A virus
Applications ELISA, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone 818
Conjugate Unconjugated
Formulation In 10mM PBS, pH 7.2 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified hepatitis A virus, Strain CF53.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific to HAV. Recognizes a neutralizing epitope on VP3.
Target Species Virus
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.