Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B virus surface (ad/ay), Mouse, Clone: BID40, Abnova™

Catalog No. 89135603 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-603 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-603 Supplier Abnova Corporation Supplier No. MAB4070
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against hepatitis B virus surface antigen (ad/ay).

Specifications

Antigen Hepatitis B virus surface (ad/ay)
Applications ELISA, Radioimmune Assays (RIA)
Classification Monoclonal
Clone BID40
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against hepatitis B virus surface antigen (ad/ay).
Dilution The optimal working dilution should be determined by the end user.
Formulation In 15mM potassium phosphate buffer, 0.85% NaCl, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Hepatitis B surface antigen (HBsAg).
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with the ad and ay subtype of the Hepatitis B virus surface antigen.
Target Species Virus
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.