Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B virus surface antigen, Mouse, Clone: 17-1-5, Abnova™

Catalog No. 89135017 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-017 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-017 Supplier Abnova Corporation Supplier No. MAB1692

Mouse monoclonal antibody raised against full-length recombinant hepatitis B virus surface antigen.

Sequence: virus

Specifications

Antigen Hepatitis B virus surface antigen
Applications ELISA
Classification Monoclonal
Clone 17-1-5
Conjugate Unconjugated
Formulation In 10mM phosphate buffer, 137mM NaCl, 2.7mM KCl, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Recombinant protein corresponding to full length hepatitis B virus surface antigen.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.