Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B virus surface, Mouse, Clone: 86C, Abnova™

Catalog No. 89134944 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-944 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-944 Supplier Abnova Corporation Supplier No. MAB1349
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native hepatitis B virus surface antigen.

Specifications

Antigen Hepatitis B virus surface
Applications ELISA
Classification Monoclonal
Clone 86C
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native hepatitis B virus surface antigen.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Host Species Mouse
Immunogen Native purified hepatitis B virus (Ad/Ay) surface Ag.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Immunoreactive with all HBsAg A antigens. Captures The HBsAg antigen and may be paired with 67A monoclonal antibody, catalog# MAB1348.
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.