Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B surface antigen, Mouse, Clone: S1-210, Abnova™

Catalog No. 89135935 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-935 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-935 Supplier Abnova Corporation Supplier No. MAB5317

Mouse monoclonal antibody raised against hepatitis B virus surface antigen.

Specifications

Antigen Hepatitis B surface antigen
Applications Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone S1-210
Conjugate Unconjugated
Formulation In PBS (0.09% sodium azide, 1% BSA)
Host Species Mouse
Immunogen Hepatitis B surface antigen.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.