Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hepatitis C virus 1b (HCV), envelope glycoprotein E2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89137528 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-528 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-528 Supplier Abnova Corporation Supplier No. PAB1272A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against hepatitis C virus 1b (HCV), envelope glycoprotein E2.

Specifications

Antigen hepatitis C virus 1b (HCV), envelope glycoprotein E2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against hepatitis C virus 1b (HCV), envelope glycoprotein E2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol)
Host Species Mouse
Immunogen Recombinant Fc fusion protein corresponding to amino acids 412-661 of Hepatitis C virus 1b (HCV), E2 protein lacking hypervariable region 1 (HVR1).
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.