Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis C virus core protein, Mouse, Clone: A1/3D1, Abnova™

Catalog No. 89136158 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-158 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-158 Supplier Abnova Corporation Supplier No. MAB6324
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of hepatitis C virus core protein.

Specifications

Antigen Hepatitis C virus core protein
Applications ELISA
Classification Monoclonal
Clone A1/3D1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of hepatitis C virus core protein.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS, pH 7.2
Host Species Mouse
Immunogen A synthetic peptide corresponding to Human hepatitis C virus (HCV) core Protein.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is reactive with recombinant CPC (Capsid Protein C)+EPM (Envelope Protein M) (core) (amino acids 1-142) and Synthetic CPC (amino acids 1-61). This clone recognizes different antigenic determinants of HCV capsid protein.
Target Species Human
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.