Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis C virus NS-3, Mouse, Clone: 24-8, Abnova™

Catalog No. 89136290 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-290 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-290 Supplier Abnova Corporation Supplier No. MAB6773
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant Hepatitis C virus NS-3.

Specifications

Antigen Hepatitis C virus NS-3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 24-8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Hepatitis C virus NS-3.
Dilution Western Blot (4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS, pH 7.2
Host Species Mouse
Immunogen Recombinant protein corresponding to least 60 residues in length of Chinese HCV NS-3.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody has no cross reaction with HCV capsid region and other non-structural regions can be seen.
Target Species Human
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.