Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis C virus NS-3, Mouse, Clone: 281, Abnova™

Catalog No. 89136165 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-165 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-165 Supplier Abnova Corporation Supplier No. MAB6331
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant hepatitis C virus NS-3.

Specifications

Antigen Hepatitis C virus NS-3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 281
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant hepatitis C virus NS-3.
Dilution Western Blot (0.1-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS, pH 7.2
Host Species Mouse
Immunogen Recombinant protein corresponding to Human hepatitis C virus (HCV) NS-3.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with recombinant NS-3 (amino acids 1252-1477), synthetic NS-3 (amino acids 1378-1458), and recombinant Chimeric HCV Polyprotein (60 KDa).
Target Species Human, Virus
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.