Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis C virus NS3 antigen, Mouse, Clone: 9-G2, Abnova™

Catalog No. 89134677 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-677 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-677 Supplier Abnova Corporation Supplier No. MAB0488

Mouse monoclonal antibody raised against recombinant hepatitis C virus NS3 antigen.

Specifications

Antigen Hepatitis C virus NS3 antigen
Applications ELISA, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 9-G2
Conjugate Unconjugated
Formulation In PBS, pH 7.2 (0.01% sodium azide)
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1350-1460 of hepatitis C virus NS3 antigen.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.