Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis C virus NS4B, Mouse, Clone: NS4-2, Abnova™

Catalog No. 89109053 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-053 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-053 Supplier Abnova Corporation Supplier No. MAB10848
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant Hepatitis C virus NS4B.

Specifications

Antigen Hepatitis C virus NS4B
Applications ELISA, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone NS4-2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Hepatitis C virus NS4B.
Dilution ELISA (1:1000000) Western Blot (1:1000) Immunohistochemistry (Frozen sections) (1:20) The optimal working dilution should be determined by the end user.
Formulation In 1X PBS, pH 7.2 (0.01% sodium azide)
Gene HCVgp1
Gene Symbols HCVgp1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1713-1717 of Hepatitis C virus NS4B.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 951475
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.