Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis Virus gp1, Mouse, Clone: 2C6.H1, Abnova™

Catalog No. 89110748 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-748 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-748 Supplier Abnova Corporation Supplier No. MAB1994
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant MHVgp1.

Specifications

Antigen Hepatitis Virus gp1
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 2C6.H1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant MHVgp1.
Dilution Western Blot (1:1000) Immunofluorescence (1:1000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene 1
Gene Symbols 1
Host Species Mouse
Immunogen Recombinant protein corresponding to full length mouse Hepatitis Virus (strain A59) MHVgp1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1489749
Test Specificity This antibody is directed against the MHV-A59 nsp9 protein. No cross reactivity occurs with SARS CoV nsp9.
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.