Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatocyte, Mouse, Clone: OCH1E5, Abnova™

Catalog No. 89134857 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-857 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-857 Supplier Abnova Corporation Supplier No. MAB11357
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against hepatocyte.

Specifications

Antigen Hepatocyte
Applications Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone OCH1E5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against hepatocyte.
Dilution Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (0.5-1 ug/mL for 30 min at RT) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.05% BSA, 0.05% sodium azide)
Host Species Mouse
Immunogen Extract of a formalin-fixed, rejected-allograft of a human liver.
Purification Method Protein A/G affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Canine, Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.