Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes simplex virus type 1, Mouse, Clone: 78G5, Abnova™

Catalog No. 89136663 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-663 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-663 Supplier Abnova Corporation Supplier No. MAB8970
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of Herpes simplex virus type 1.

Specifications

Antigen Herpes simplex virus type 1
Applications Immunofluorescence
Classification Monoclonal
Clone 78G5
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of Herpes simplex virus type 1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.02% sodium azide)
Host Species Mouse
Immunogen A synthetic peptide corresponding to Herpes simplex virus type 1.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Herpes Simplex virus (HSV)
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.