Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes simplex virus type 1 (gC), Mouse, Clone: T96, Abnova™

Catalog No. 89135493 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-493 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-493 Supplier Abnova Corporation Supplier No. MAB3654
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant herpes simplex virus type 1 (gC).

Specifications

Antigen Herpes simplex virus type 1 (gC)
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone T96
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant herpes simplex virus type 1 (gC).
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Recombinant protein corresponding to Herpes simplex virus type 1 (gC).
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes gC antigen of HSV1 (Herpes Simplex Virus type 1), a dsDNA virus, member of Herpesviridae family.
Target Species Virus
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.