Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes simplex virus type 2 Glycoprotein D, Mouse, Clone: 910, Abnova™

Catalog No. 89135800 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-800 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-800 Supplier Abnova Corporation Supplier No. MAB4802

Mouse monoclonal antibody raised against herpes simplex virus type 2 Glycoprotein D.

Specifications

Antigen Herpes simplex virus type 2 Glycoprotein D
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 910
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against herpes simplex virus type 2 Glycoprotein D.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified Herpes simplex virus type 2 virions.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity HSV2, specific to gD.
Target Species Herpes Simplex virus (HSV)
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Herpes simplex virus type 2 Glycoprotein D monoclonal antibody, clone 910
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.