Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Herpes virus type 6 gp90, Mouse, Clone: 300, Abnova™
Mouse monoclonal antibody raised against human herpes virus type 6 gp90.
Supplier: Abnova Corporation MAB4806
Specifications
Herpes virus type 6 gp90 | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Mouse | |
1 mg | |
Primary | |
Virus | |
Liquid |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
300 | |
Mouse monoclonal antibody raised against human herpes virus type 6 gp90. | |
In 10mM PBS, pH 7.2 (0.09% sodium azide) | |
Human herpes virus type 6. | |
RUO | |
Specific for the 90KDa glycoprotein of HHV-6, Strains A and B. Not suitable for use in IFA. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction