Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes virus type 8, Mouse, Clone: 13B10, Abnova™

Catalog No. 89136399 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-399 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-399 Supplier Abnova Corporation Supplier No. MAB7925

Mouse monoclonal antibody raised against human herpesvirus type 8.

Specifications

Antigen Herpes virus type 8
Applications Immunohistochemistry
Classification Monoclonal
Clone 13B10
Concentration 30 ug/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human herpesvirus type 8.
Dilution Immunohistochemistry (1:25-1:50) The optimal working dilution should be determined by the end user.
Formulation Liquid concentrate in cell culture supernant (BSA, 0.095% sodium azide)
Host Species Mouse
Immunogen Native purified human herpesvirus type 8 (Latent Nuclear Antigen).
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.