Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Mouse anti-hIAP-1/BIRC3, Clone: 11D12.1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABC582
Description
Specifically detects hIAP-1/BIRC3 clone: 11D12.1 in Human samples, and it is validated for Western Blotting
Baculoviral IAP repeat-containing protein 3 (UniProt Q13489; also known as API2, Apoptosis inhibitor 2, Baculoviral IAP repeat-containing 3, c-IAP2, hIAP-1, hIAP1, IAP homolog C, IAP-1, Inhibitor of apoptosis protein 1, Mammalian IAP homolog C, RING finger protein 49, TNFR2-TRAF-signaling complex protein 1) is encoded by the BIRC3 (also known as API2, AIP1, IAP1, HAIP1, HIAP1, MALT2, MIHC, RNF49) gene (Gene ID 330) in human. hIAP-1 (c-IAP2) belongs to the inhibitor of apoptosis (IAP) family of proteins characterized by the presence of at least one baculoviral IAP repeat (BIR) domain. hIAP-1 (c-IAP2 contains three BIR (BIR1/2/3) domains, followed by an UBA (ubiquitin-associated), a CARD (caspase activation and recruitment domain), and a RING (really interesting new gene) domain. The BIR domains mediate hIAP-1 interaction with various binding partners, such as caspases, other IAP family members, and receptor-associated complexes. The RING domain and its associated E3 ligase mediates not only the ubiqutination of substrate proteins, such as caspase-3, caspase-7, SMAC/DIABLO, but also self-ubiquitylation. The UBA domain enables hIAP-1 to engage protein complexes containing poly-ubiquitylated protein(s).
Specifications
hIAP-1/BIRC3 | |
Monoclonal | |
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
BIRC3; API2; IAP1; MIHC; RNF49 | |
GST-tagged recombinant protein corresponding to the BIR 1/2 of human hIAP-1/BIRC3. | |
100 μg | |
Apoptosis, Cancer | |
NP_001156 | |
Stable for one year at 2°C to 8°C from date of receipt. | |
IgG1 κ |
Western Blot | |
11D12.1 | |
Q13489 | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction