Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIP1, Mouse, Clone: 1B11, Abnova™

Catalog No. 89110944 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-944 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-944 Supplier Abnova Corporation Supplier No. MAB2341
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against HIP1.

The product of this gene is a membrane-associated protein that colocalizes with huntingtin. This protein has similarities to cytoskeleton proteins and its interaction with huntingtin is thought to play a functional role in the cell filament network. Loss of normal huntingtin-HIP1 interaction in Huntington disease may contribute to a defect in membrane-cytoskeletal integrity in the brain. This gene could help in the understanding of the normal function of huntingtin and also the pathogenesis of Huntington disease. It also has been implicated in the pathogenesis of hematopoietic malignancies. An alternative splice variant of this gene has been described but its full length sequence has not been determined. [provided by RefSeq

Specifications

Antigen HIP1
Applications Western Blot
Classification Monoclonal
Clone 1B11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against HIP1.
Dilution Western Blot (1:10000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Gene HIP1
Gene Alias ILWEQ/MGC126506
Gene Symbols HIP1
Host Species Mouse
Immunogen Human 3'-HIP1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3092
Test Specificity This antibody is specific to human HIP1. Does not cross-react with HIP1r.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.