Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIP1R, Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89112758 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-758 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-758 Supplier Abnova Corporation Supplier No. MAB6239
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against HIP1R.

Specifications

Antigen HIP1R
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against HIP1R.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.09% sodium azide)
Gene HIP1R
Gene Alias FLJ14000/FLJ27022/HIP12/HIP3/ILWEQ/KIAA0655/MGC47513
Gene Symbols HIP1R
Host Species Mouse
Immunogen Human HIP1r-TALIN Homology Region (57 kDa).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9026
Test Specificity This is specific to HIP1r.
Target Species Human, Rat
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.