Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIST2H3A, Mouse, Clone: 6D10H9, Abnova™

Catalog No. 89113486 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-486 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-486 Supplier Abnova Corporation Supplier No. MAB8548

Mouse monoclonal antibody raised against synthetic peptide of HIST2H3A.

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. This structure consists of approximately 146bp of DNA wrapped around a nucleosome, an octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H3 family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. This gene is found in a histone cluster on chromosome 1. This gene is one of four histone genes in the cluster that are duplicated; this record represents the centromeric copy. [provided by RefSeq

Specifications

Antigen HIST2H3A
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6D10H9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of HIST2H3A.
Dilution ELISA (1:5000-1:8000) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS, pH 7.5, 0.1% BSA
Gene HIST2H3A
Gene Alias H3/n/H3/o
Gene Symbols HIST2H3A
Host Species Mouse
Immunogen A synthetic peptide corresponding to human HIST2H3A.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 333932
Test Specificity This antibody is not only able to bind to H3R2a, but also distinguish two forms (Asymmetric and Symmetric) of H3R2. It specifically detected protein derived from cell lysates in both ELISA and WB.
Target Species Human
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.