Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-HIV-1 p24, HRP, Clone: 8G9, Novus Biologicals™

Supplier: Novus Biologicals NBP241338
Specifications
HIV-1 p24 | |
Monoclonal | |
1 mg/mL | |
Western Blot 0.2- 0.5ug/mL, ELISA 0.2- 0.5ug/mL | |
Capsid protein p24, Human immunodeficiency virus 1, Human immunodeficiency virus type 1 p24 | |
Antibody was raised against a recombinant full-length HIV-1 p24 protein. | |
Protein A purified | |
RUO | |
155348 | |
Virus | |
Purified |
Western Blot, ELISA | |
8G9 | |
HRP | |
PBS with 0.02% Thimerosal | |
Mouse | |
56 kDa | |
0.1 mg | |
Primary | |
By Western blot, anti-HIV-1 p24 antibody detects a ∼24 kDa, a ∼41 kDa, and a ∼55 kDa protein, corresponding to HIV-1 p24 and to its precursors p41 and p55, respectively, in HIV-1 samples. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction