Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-A/HLA-B/HLA-C, Mouse, Clone: LY5.1, Abnova™

Catalog No. 89112843 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-843 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-843 Supplier Abnova Corporation Supplier No. MAB6383

Mouse monoclonal antibody raised against HLA-A/HLA-B/HLA-C.

Specifications

Antigen HLA-A/HLA-B/HLA-C
Applications Immunohistochemistry
Classification Monoclonal
Clone LY5.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against HLA-A/HLA-B/HLA-C.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2
Gene Alias HLAA
Gene Symbols HLA-A
Host Species Mouse
Immunogen Purified human HLA-A/HLA-B/HLA-C complex.
Purification Method Protein G purification
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3105/3106/3107
Target Species Human
Content And Storage Store at -20°C.
After reconstitution with deionized water, store at -20°C or lower.
Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.