Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-ABC Mouse, BUV395, Clone: G46-2.6, BD

Catalog No. BDB565334 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565334 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565334 Supplier BD Biosciences Supplier No. 565334
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Human Leukocyte Antigen (HLA) complex is the human version of the MHC, helping the immune system distinguish the body's own proteins versus those from foreign invaders, such as viruses. Humans have three main MHC class I genes, known as HLA-A, HLA-B and HLA-C. Major histocompatibility complex (MHC) class I molecules, which are widely found on the surface of nucleated cells, function by binding peptides and displaying them on the cell surface to cytotoxic T-cells. Intracellular degradation of cytosolic proteins by the proteasome generates many of the peptides that load MHC class I molecules. MHC class I may also serve as an inhibitory ligand for natural killer (NK) cell receptors (KIR, Killer Immunoglobulin-like Receptors), which viruses may modulate expression levels for to evade immune detection. The G46-2.6 monoclonal antibody binds to a monomorphic epitope on the alpha chain of HLA-A, HLA-B and HLA-C.

The antibody was conjugated to BD Horizon BUV395 which has been exclusively developed by BD Biosciences as an optimal dye for use on a 355nm laser equipped instrument. With an Ex Max at 348nm and an Em Max at 395nm, this dye has virtually no spillover into any other detector. BD Horizon BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.

TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-ABC
Applications Flow Cytometry
Classification Monoclonal
Clone G46-2.6
Conjugate Brilliant Ultraviolet 395
Description Major histocompatibility complex, class I, A,B,C; HLA class I A,B,C
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Human, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.