Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-B, Mouse, Clone: EP-4, Abnova™

Catalog No. 89112164 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-164 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-164 Supplier Abnova Corporation Supplier No. MAB5202
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native HLA-B.

HLA-B belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. Class I molecules play a central role in the immune system by presenting peptides derived from the endoplasmic reticulum lumen. They are expressed in nearly all cells. The heavy chain is approximately 45kDa and its gene contains 8 exons. Exon 1 encodes the leader peptide, exon 2 and 3 encode the alpha1 and alpha2 domains, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region and exons 6 and 7 encode the cytoplasmic tail. Polymorphisms within exon 2 and exon 3 are responsible for the peptide binding specificity of each class one molecule. Typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. Hundreds of HLA-B alleles have been described. [provided by RefSeq

Specifications

Antigen HLA-B
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone EP-4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native HLA-B.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene HLA-B
Gene Alias AS/HLA-B-7301/HLA-B73/HLAB/HLAC/SPDA1
Gene Symbols HLA-B
Host Species Mouse
Immunogen Native purified from lymphocytes of HLA-B27 individual.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3106
Test Specificity EP-4 recognizes the HLA-B27 cell surface antigen on human cells. Since EP-4 recognizes the HLA-B27 antigen, it may be used to HLA type human lymphoctyes. Approximately 60% of patients with ankylosing spondylitis are HLA-B27 positive. This reagent can be used to help identify this HLA haplotype in human lymphocytes. Monoclonal antibody EP-4 recognizes the HLA-B27 antigen expressed on individuals with this HLA haplotype.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.