Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-Class I, Mouse, Biotin, Clone: W6/32, Abnova™

Catalog No. 89135897 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-897 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-897 Supplier Abnova Corporation Supplier No. MAB5082
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native HLA-Class I.

Specifications

Antigen HLA-Class I
Applications Flow Cytometry
Classification Monoclonal
Clone W6/32
Conjugate Biotin
Description Mouse monoclonal antibody raised against native HLA-Class I.
Dilution Flow Cytometry (1:300) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified HLA-Class I from membrane of human tonsil cells.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes MHC Class I molecules (MHC Class Ia) that are expressed on the surface of all human nucleated cell types. This antibody is also suitable as a positive control for HLA tissue typing and crossmatching.
Target Species Bovine, Feline, Human, Primate
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.