Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA Class I, Mouse, Clone: MEM-123, Abnova™

Catalog No. 89111635 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-635 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-635 Supplier Abnova Corporation Supplier No. MAB3854
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native HLA Class I.

Specifications

Antigen HLA Class I
Applications ELISA, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone MEM-123
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native HLA Class I.
Dilution The optimal working dilution should be determined by the end user.
Formulation In TBS, pH 8.0 (0.09% sodium azide)
Gene Alias HLAA
Gene Symbols HLA-A
Host Species Mouse
Immunogen Native purified HLA Class I from African Green Monkey COS-7 cells.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3105/3133/3134/3135
Test Specificity This antibody reacts with all human classical MHC Class I molecules (majorhistocompatibility complex) in native cell-surface forms as well as with human HLA-G cDNA transfected cells. MHC Class I molecules (MHC Class Ia) are expressed on the surface ofall human cell types.This antibody completely blocks binding of classical W6/32 to surface-expressed HLA-G, but does not cross-blocks this antibody.
Target Species Bovine, Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.