Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-Class I, Mouse, Clone: MEM-81, Abnova™

Catalog No. 89108232 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-232 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-232 Supplier Abnova Corporation Supplier No. MAB0904
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native HLA-Class I.

Specifications

Antigen HLA-Class I
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-81
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native HLA-Class I.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene Alias HLAA
Gene Symbols HLA-A
Host Species Mouse
Immunogen Native purified HLA-Class I from leukocytes of a patient suffering from a LGL-type leukemia.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3105/3106/3107
Test Specificity This antibody reacts with human classical MHC Class I molecules in native cell-surface forms.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.