Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-DR1, Mouse, Clone: MEM-267, Abnova™

Catalog No. 89135203 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-203 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-203 Supplier Abnova Corporation Supplier No. MAB2527
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant HLA-DR1.

Specifications

Antigen HLA-DR1
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone MEM-267
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant HLA-DR1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids of human HLA-DR1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody MEM-267 specifically binds to the empty but not peptide-loaded form of HLA-DR1.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.