Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-E, Mouse, Clone: MEM-E/08, Abnova™

Catalog No. 89111508 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-508 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-508 Supplier Abnova Corporation Supplier No. MAB3632
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant HLA-E.

HLA-E belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. HLA-E binds a restricted subset of peptides derived from the leader peptides of other class I molecules. The heavy chain is approximately 45kDa and its gene contains 8 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domains, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exons 6 and 7 encode the cytoplasmic tail. [provided by RefSeq

Specifications

Antigen HLA-E
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone MEM-E/08
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant HLA-E.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene HLA-E
Gene Alias DKFZp686P19218/EA1.2/EA2.1/HLA-6.2/MHC/QA1
Gene Symbols HLA-E
Host Species Mouse
Immunogen Recombinant protein corresponding to human HLA-E.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3133
Test Specificity This antibody recognized native surface-expressed HLA-E, but not denaturated heavy chain of HLA-E. HLA-E belongs to the MHC Class I molecules (MHC Class Ib; nonclassical) and it is expressed on many types of the human cells. The published results revealed that antibody is remarkably specific to HLA-E, only with weak cross-reactivity with following classical MHC Class I molecules (MHC Class Ia) : HLA-A24, -B7, -B27, -B51, -B54, -C7.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.