Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-G, Mouse, Clone: F21-21, Abnova™

Catalog No. 89112746 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-746 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-746 Supplier Abnova Corporation Supplier No. MAB6225
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native HLA-G.

Specifications

Antigen HLA-G
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone F21-21
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native HLA-G.
Dilution Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Gene HLA-G
Gene Symbols HLA-G
Host Species Mouse
Immunogen Native purified from white blood cells from chicken plasma.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 768350
Test Specificity The beta2-microglobulin subunit (Mr ∼ 14 KDa) of avian MHC Class I molecules.
Target Species Chicken
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.